Search
Search

jupa| Gene Symbol: jupa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131177.1 | NP_571252.1 | 15 - 16 | 2318 | 78 | |
| GenBank mRNA | AF099738.1 | - | 15 - 16 | 2318 | 78 | |
| BC058305.1 | - | 15 - 16 | 2339 | 78 | ||
Set Membership: |
|