Search
Search

notch1b| Gene Symbol: notch1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131302.2 | NP_571377.2 | 25 - 26 | 4656 | 80 | |
| XM_005168649.4 | XP_005168706.1 | 25 - 26 | 4804 | 80 | ||
| GenBank mRNA | AY805407.1 | - | 25 - 26 | 4656 | 80 | |
Set Membership: |
|