Search
Search

cldn11b| Gene Symbol: cldn11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131772.2 | NP_571847.2 | 2 - 3 | 557 | 90 | |
| GenBank mRNA | AF359429.1 | - | 2 - 3 | 549 | 90 | |
| BC139523.1 | - | 2 - 3 | 557 | 90 | ||
Set Membership: |
|