Search
Search

thbs4b| Gene Symbol: thbs4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173226.1 | NP_775333.1 | 8 - 9 | 1218 | 73 | |
| GenBank mRNA | AY081009.1 | - | 8 - 9 | 1218 | 73 | |
| BC095593.1 | - | 8 - 9 | 1228 | 73 | ||
Set Membership: |
|