Search
Search

anxa11a| Gene Symbol: anxa11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181765.2 | NP_861430.2 | 12 - 13 | 1419 | 74 | |
| NM_183410.2 | NP_899670.1 | 12 - 13 | 1290 | 74 | ||
| GenBank mRNA | AY178801.1 | - | 12 - 13 | 1411 | 74 | |
| BC053208.1 | - | 12 - 13 | 1252 | 74 | ||
Set Membership: |
|