Search
Search

krit1| Gene Symbol: krit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317001.1 | NP_001303930.1 | 2 - 3 | 195 | 94 | |
| GenBank mRNA | BC055149.1 | - | 2 - 3 | 180 | 94 | |
| EB920540.1 | - | 2 - 3 | 195 | 94 | ||
Set Membership: |
|