Search
Search

tomm40| Gene Symbol: tomm40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199614.1 | NP_955908.1 | 4 - 5 | 404 | 70 | |
| XM_005158091.4 | XP_005158148.1 | 4 - 5 | 812 | 70 | ||
| XM_017351318.2 | XP_017206807.1 | 4 - 5 | 601 | 70 | ||
| GenBank mRNA | BC053295.1 | - | 4 - 5 | 404 | 70 | |
Set Membership: |
|