Search
Search

rem1| Gene Symbol: rem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201174.1 | NP_957468.1 | 2 - 3 | 450 | 132 | |
| XM_009306022.3 | XP_009304297.1 | 2 - 3 | 1049 | 132 | ||
| GenBank mRNA | BC045943.1 | - | 2 - 3 | 450 | 132 | |
| BC165381.1 | - | 1 - 2 | 364 | 132 | ||
| KJ826439.1 | - | 1 - 2 | 132 | |||
Set Membership: |
|