Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03090285_m1
          See other MYCBP2 GE Assays ›
          Gene Symbol:
          mycbp2
          Gene Name:
          MYC binding protein 2
          Gene Aliases:
          Myc, PAM, esr, esrom, fa97c06, ns:zft368, si:busm1-12g7.1, si:dz12g7.1, si:dz256p4.1, til, tilsit, wu:fa97c06, xx:zft368, zft368
          Chromosome Location:
          Chr.9: 220341 - 485433 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03090285_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: mycbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012247.2 NP_001012247.2 68 - 69 11996 77
          XM_009304548.3 XP_009302823.1 72 - 73 12589 77
          XM_009304549.3 XP_009302824.1 72 - 73 12586 77
          XM_009304550.3 XP_009302825.1 72 - 73 12565 77
          XM_009304551.3 XP_009302826.1 71 - 72 12514 77
          XM_009304552.3 XP_009302827.1 71 - 72 12490 77
          XM_009304553.3 XP_009302828.1 71 - 72 12487 77
          XM_009304554.3 XP_009302829.1 71 - 72 12484 77
          XM_009304555.3 XP_009302830.1 72 - 73 12481 77
          XM_009304556.3 XP_009302831.1 71 - 72 12460 77
          XM_009304557.3 XP_009302832.1 71 - 72 12451 77
          XM_009304558.3 XP_009302833.1 71 - 72 12424 77
          XM_009304559.3 XP_009302834.1 70 - 71 12325 77
          XM_009304560.3 XP_009302835.1 70 - 71 12292 77
          XM_009304561.3 XP_009302836.1 70 - 71 12286 77
          XM_009304562.3 XP_009302837.1 69 - 70 12256 77
          XM_009304563.3 XP_009302838.1 69 - 70 12250 77
          XM_017357656.2 XP_017213145.1 72 - 73 12550 77
          XM_021478618.1 XP_021334293.1 71 - 72 12406 77
          GenBank mRNA AY818192.1 - 68 - 69 11861 77

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Danio rerio Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          axon guidance
          axon choice point recognition
          protein ubiquitination
          central nervous system projection neuron axonogenesis
          habenula development
          retinal ganglion cell axon guidance
          circadian regulation of gene expression
          regulation of pteridine metabolic process
          developmental pigmentation
          rhythmic process
          neuromuscular process
          regulation of axon guidance
          ubiquitin-protein transferase activity
          guanyl-nucleotide exchange factor activity
          protein binding
          Ran GTPase binding
          transferase activity
          metal ion binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline