Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dr03090287_m1
          See other MYCBP2 GE Assays ›
          Gene Symbol:
          mycbp2
          Gene Name:
          MYC binding protein 2
          Gene Aliases:
          Myc, PAM, esr, esrom, fa97c06, ns:zft368, si:busm1-12g7.1, si:dz12g7.1, si:dz256p4.1, til, tilsit, wu:fa97c06, xx:zft368, zft368
          Chromosome Location:
          Chr.9: 220341 - 485433 on Build GRCz11
          Species:
          Zebrafish
          Species Specific ID (Flybase ID):
          -
          Assay ID Dr03090287_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: mycbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012247.2 NP_001012247.2 70 - 71 12549 73
          XM_009304548.3 XP_009302823.1 74 - 75 13142 73
          XM_009304549.3 XP_009302824.1 74 - 75 13139 73
          XM_009304550.3 XP_009302825.1 74 - 75 13118 73
          XM_009304551.3 XP_009302826.1 73 - 74 13067 73
          XM_009304552.3 XP_009302827.1 73 - 74 13043 73
          XM_009304553.3 XP_009302828.1 73 - 74 13040 73
          XM_009304554.3 XP_009302829.1 73 - 74 13037 73
          XM_009304555.3 XP_009302830.1 74 - 75 13034 73
          XM_009304556.3 XP_009302831.1 73 - 74 13013 73
          XM_009304557.3 XP_009302832.1 73 - 74 13004 73
          XM_009304558.3 XP_009302833.1 73 - 74 12977 73
          XM_009304559.3 XP_009302834.1 72 - 73 12878 73
          XM_009304560.3 XP_009302835.1 72 - 73 12845 73
          XM_009304561.3 XP_009302836.1 72 - 73 12839 73
          XM_009304562.3 XP_009302837.1 71 - 72 12809 73
          XM_009304563.3 XP_009302838.1 71 - 72 12803 73
          XM_017357656.2 XP_017213145.1 74 - 75 13103 73
          XM_021478618.1 XP_021334293.1 73 - 74 12959 73
          GenBank mRNA AY818192.1 - 70 - 71 12414 73

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Danio rerio Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          axon guidance
          axon choice point recognition
          protein ubiquitination
          central nervous system projection neuron axonogenesis
          habenula development
          retinal ganglion cell axon guidance
          circadian regulation of gene expression
          regulation of pteridine metabolic process
          developmental pigmentation
          rhythmic process
          neuromuscular process
          regulation of axon guidance
          ubiquitin-protein transferase activity
          guanyl-nucleotide exchange factor activity
          protein binding
          Ran GTPase binding
          transferase activity
          metal ion binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline