Search
Search

tsn| Gene Symbol: tsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025452.1 | NP_001020623.1 | 5 - 6 | 655 | 73 | |
| GenBank mRNA | BC096804.1 | - | 5 - 6 | 655 | 73 | |
| BC109403.1 | - | 5 - 6 | 601 | 73 | ||
| BC165426.1 | - | 5 - 6 | 490 | 73 | ||
Set Membership: |
|