Search
Search

setdb1b| Gene Symbol: setdb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077277.3 | NP_001070745.3 | 19 - 20 | 3417 | 110 | |
| XM_005158013.4 | XP_005158070.2 | 21 - 22 | 3628 | 110 | ||
| XM_005158015.4 | XP_005158072.2 | 21 - 22 | 3580 | 110 | ||
| XM_005158016.4 | XP_005158073.2 | 19 - 20 | 3427 | 110 | ||
| GenBank mRNA | BC124601.1 | - | 19 - 20 | 3359 | 110 | |
| DQ358103.1 | - | 12 - 13 | 2056 | 110 | ||
Set Membership: |
|