Search
Search

desmb| Gene Symbol: desmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077452.1 | NP_001070920.1 | 5 - 6 | 1065 | 72 | |
| XM_021476935.1 | XP_021332610.1 | 5 - 6 | 1087 | 72 | ||
| GenBank mRNA | BC124512.1 | - | 5 - 6 | 1065 | 72 | |
| BC165598.1 | - | 5 - 6 | 1060 | 72 | ||
Set Membership: |
|