Search
Search

epd| Gene Symbol: epd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131005.2 | NP_571080.1 | 1 - 2 | 121 | 109 | |
| GenBank mRNA | BC092712.1 | - | 1 - 2 | 121 | 109 | |
| X52502.1 | - | 91 | 109 | |||
Set Membership: |
|