Search
Search

wt1a| Gene Symbol: wt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131046.2 | NP_571121.1 | 4 - 5 | 1022 | 97 | |
| GenBank mRNA | AF144550.1 | - | 4 - 5 | 771 | 97 | |
| AY028627.1 | - | 4 - 5 | 633 | 97 | ||
| BC162349.1 | - | 4 - 5 | 731 | 97 | ||
| BC162638.1 | - | 4 - 5 | 731 | 97 | ||
Set Membership: |
|