Search
Search

mpped2| Gene Symbol: mpped2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200042.1 | NP_956336.1 | 6 - 7 | 1132 | 67 | |
| XM_005174308.4 | XP_005174365.1 | 6 - 7 | 67 | |||
| GenBank mRNA | AY639016.1 | - | 5 - 6 | 876 | 67 | |
| BC049341.1 | - | 6 - 7 | 1132 | 67 | ||
| BC153488.1 | - | 6 - 7 | 67 | |||
Set Membership: |
|