Search
Search

acsl4a| Gene Symbol: acsl4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200649.1 | NP_956943.1 | 2 - 3 | 548 | 103 | |
| GenBank mRNA | BC057476.1 | - | 2 - 3 | 548 | 103 | |
| BC164569.1 | - | 1 - 2 | 372 | 103 | ||
Set Membership: |
|