Search
Search

guca1e| Gene Symbol: guca1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200656.1 | NP_956950.1 | 2 - 3 | 378 | 57 | |
| GenBank mRNA | AY850385.1 | - | 2 - 3 | 353 | 57 | |
| BC057493.1 | - | 2 - 3 | 378 | 57 | ||
Set Membership: |
|