Search
Search

hif1an| Gene Symbol: hif1an | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201496.1 | NP_958904.1 | 7 - 8 | 1182 | 115 | |
| GenBank mRNA | BC044475.1 | - | 7 - 8 | 1182 | 115 | |
| BC164149.1 | - | 7 - 8 | 1016 | 115 | ||
Set Membership: |
|