Search
Search

tcea3| Gene Symbol: tcea3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205683.1 | NP_991246.1 | 5 - 6 | 561 | 84 | |
| XM_009292450.3 | XP_009290725.1 | 7 - 8 | 856 | 84 | ||
| GenBank mRNA | BC065444.1 | - | 5 - 6 | 561 | 84 | |
Set Membership: |
|