Search
Search

rasgef1bb| Gene Symbol: rasgef1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003479.1 | NP_001003479.1 | 6 - 7 | 971 | 72 | |
| GenBank mRNA | BC078340.1 | - | 6 - 7 | 971 | 72 | |
Set Membership: |
|