Search
Search

aqp4| Gene Symbol: aqp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003749.4 | NP_001003749.1 | 4 - 5 | 730 | 102 | |
| NM_001358313.1 | NP_001345242.1 | 4 - 5 | 730 | 102 | ||
| XM_009294620.3 | XP_009292895.1 | 4 - 5 | 888 | 102 | ||
| GenBank mRNA | BC078213.1 | - | 4 - 5 | 717 | 102 | |
| FJ666327.1 | - | 4 - 5 | 717 | 102 | ||
Set Membership: |
|