Search
Search

gspt1| Gene Symbol: gspt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003992.1 | NP_001003992.1 | 10 - 11 | 1381 | 95 | |
| GenBank mRNA | BC080263.1 | - | 10 - 11 | 1381 | 95 | |
| BC164587.1 | - | 10 - 11 | 1247 | 95 | ||
Set Membership: |
|