Search
Search

gspt1| Gene Symbol: gspt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003992.1 | NP_001003992.1 | 11 - 12 | 1551 | 94 | |
| GenBank mRNA | BC080263.1 | - | 11 - 12 | 1551 | 94 | |
| BC164587.1 | - | 11 - 12 | 1417 | 94 | ||
Set Membership: |
|