Search
Search

nudt3b| Gene Symbol: nudt3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006034.2 | NP_001006034.1 | 1 - 2 | 258 | 85 | |
| XM_017356473.2 | XP_017211962.1 | 1 - 2 | 275 | 85 | ||
| GenBank mRNA | BC083382.1 | - | 1 - 2 | 259 | 85 | |
Set Membership: |
|