Search
Search

popdc2| Gene Symbol: popdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020800.1 | NP_001018636.1 | 3 - 4 | 1458 | 76 | |
| GenBank mRNA | AY293119.1 | - | 3 - 4 | 1458 | 76 | |
| BC129264.1 | - | 3 - 4 | 1458 | 76 | ||
Set Membership: |
|