Search
Search

snx9b| Gene Symbol: snx9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030124.1 | NP_001025295.1 | 3 - 4 | 457 | 71 | |
| XM_005170181.4 | XP_005170238.1 | 3 - 4 | 477 | 71 | ||
| XM_021468448.1 | XP_021324123.1 | 3 - 4 | 477 | 71 | ||
| XM_021468449.1 | XP_021324124.1 | 3 - 4 | 477 | 71 | ||
| GenBank mRNA | BC096861.1 | - | 3 - 4 | 401 | 71 | |
| BC115194.1 | - | 3 - 4 | 460 | 71 | ||
| BC163586.1 | - | 3 - 4 | 276 | 71 | ||
| BC163590.1 | - | 3 - 4 | 276 | 71 | ||
| BC167965.1 | - | 3 - 4 | 438 | 71 | ||
Set Membership: |
|