Search
Search

nptxrb| Gene Symbol: nptxrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044314.1 | NP_001037779.1 | 4 - 5 | 1477 | 103 | |
| GenBank mRNA | BC067657.1 | - | 4 - 5 | 1351 | 103 | |
| BC109464.1 | - | 5 - 6 | 1389 | 103 | ||
| BC131861.1 | - | 4 - 5 | 1477 | 103 | ||
| BC171437.1 | - | 4 - 5 | 1466 | 103 | ||
| BC171439.1 | - | 4 - 5 | 1466 | 103 | ||
Set Membership: |
|