Search
Search

drc3| Gene Symbol: drc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077727.1 | NP_001071195.1 | 12 - 13 | 1502 | 70 | |
| XM_021471579.1 | XP_021327254.1 | 12 - 13 | 1511 | 70 | ||
| GenBank mRNA | BC125885.1 | - | 12 - 13 | 1502 | 70 | |
Set Membership: |
|