Search
Search

dla| Gene Symbol: dla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130954.2 | NP_571029.2 | 2 - 3 | 763 | 69 | |
| GenBank mRNA | AF030031.1 | - | 2 - 3 | 754 | 69 | |
| BC075742.1 | - | 2 - 3 | 763 | 69 | ||
| CU458989.1 | - | 2 - 3 | 754 | 69 | ||
| DQ993185.1 | - | 2 - 3 | 754 | 69 | ||
Set Membership: |
|