Search Thermo Fisher Scientific
Gene Symbol: col2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131292.1 | NP_571367.1 | 28 - 29 | 1940 | 75 | |
XM_005166863.4 | XP_005166920.1 | 27 - 28 | 1872 | 75 | ||
GenBank mRNA | BC163934.1 | - | 28 - 29 | 1932 | 75 | |
BC167477.1 | - | 27 - 28 | 1836 | 75 | ||
BC168504.1 | - | 28 - 29 | 2021 | 75 | ||
DQ335127.1 | - | 28 - 29 | 1940 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |