Search
Search

fosab| Gene Symbol: fosab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205569.1 | NP_991132.1 | 3 - 4 | 533 | 73 | |
| GenBank mRNA | BC065466.1 | - | 3 - 4 | 533 | 73 | |
| DQ003339.2 | - | 3 - 4 | 428 | 73 | ||
Set Membership: |
|