Search
Search

amy2a| Gene Symbol: amy2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213011.2 | NP_998176.2 | 9 - 10 | 1375 | 134 | |
| GenBank mRNA | BC054669.1 | - | 9 - 10 | 1350 | 134 | |
| BC062867.1 | - | 9 - 10 | 1352 | 134 | ||
Set Membership: |
|