Search
Search

cul1b| Gene Symbol: cul1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213495.1 | NP_998660.1 | 17 - 18 | 2161 | 119 | |
| XM_021470126.1 | XP_021325801.1 | 17 - 18 | 2248 | 119 | ||
| GenBank mRNA | BC048370.1 | - | 17 - 18 | 2161 | 119 | |
| BC165368.1 | - | 16 - 17 | 1927 | 119 | ||
Set Membership: |
|