Search
Search

sdf2l1| Gene Symbol: sdf2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003730.1 | NP_001003730.2 | 1 - 2 | 193 | 65 | |
| GenBank mRNA | BC078401.1 | - | 1 - 2 | 193 | 65 | |
| BC154283.1 | - | 1 - 2 | 225 | 65 | ||
Set Membership: |
|