Search
Search

mlf2| Gene Symbol: mlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025516.1 | NP_001020687.1 | 7 - 8 | 1024 | 78 | |
| GenBank mRNA | BC097157.1 | - | 7 - 8 | 1024 | 78 | |
| BC154139.1 | - | 7 - 8 | 1457 | 78 | ||
Set Membership: |
|