Search
Search

slc8a1a| Gene Symbol: slc8a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037102.1 | NP_001032179.1 | 3 - 4 | 1947 | 110 | |
| XM_009306112.3 | XP_009304387.1 | 4 - 5 | 2139 | 110 | ||
| XM_021479613.1 | XP_021335288.1 | 4 - 5 | 2150 | 110 | ||
| XM_021479615.1 | XP_021335290.1 | 3 - 4 | 258 | 110 | ||
| GenBank mRNA | AY934775.1 | - | 3 - 4 | 1947 | 110 | |
| BC095839.1 | - | 4 - 5 | 2047 | 110 | ||
| JN016748.1 | - | 3 - 4 | 1947 | 110 | ||
Set Membership: |
|