Search
Search

tnika| Gene Symbol: tnika | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199703.1 | NP_955997.1 | 15 - 16 | 1969 | 70 | |
| XM_005163387.4 | XP_005163444.1 | 17 - 18 | 2348 | 70 | ||
| XM_005163388.4 | XP_005163445.1 | 17 - 18 | 2348 | 70 | ||
| XM_005163389.4 | XP_005163446.1 | 16 - 17 | 2261 | 70 | ||
| XM_005163390.4 | XP_005163447.1 | 16 - 17 | 2087 | 70 | ||
| XM_005163391.4 | XP_005163448.1 | 16 - 17 | 2087 | 70 | ||
| XM_005163392.4 | XP_005163449.1 | 15 - 16 | 2000 | 70 | ||
| XM_005163393.4 | XP_005163450.1 | 2 - 3 | 473 | 70 | ||
| GenBank mRNA | BC055134.1 | - | 15 - 16 | 1969 | 70 | |
Set Membership: |
|