Search
Search

penka| Gene Symbol: penka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200083.2 | NP_956377.2 | 2 - 3 | 340 | 65 | |
| GenBank mRNA | BC059668.1 | - | 2 - 3 | 320 | 65 | |
| EH439307.1 | - | 2 - 3 | 351 | 65 | ||
Set Membership: |
|