Search
Search

myo1eb| Gene Symbol: myo1eb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200636.1 | NP_956930.1 | 25 - 26 | 3303 | 96 | |
| GenBank mRNA | BC057426.1 | - | 25 - 26 | 3303 | 96 | |
| BC164037.1 | - | 24 - 25 | 3067 | 96 | ||
Set Membership: |
|