Search
Search

bysl| Gene Symbol: bysl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201106.1 | NP_957400.1 | 1 - 2 | 307 | 91 | |
| GenBank mRNA | AY648821.1 | - | 1 - 2 | 329 | 91 | |
| BC053134.1 | - | 1 - 2 | 307 | 91 | ||
Set Membership: |
|