Search
Search

stt3a| Gene Symbol: stt3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201458.2 | NP_958866.1 | 10 - 11 | 1224 | 78 | |
| GenBank mRNA | BC046072.1 | - | 10 - 11 | 1225 | 78 | |
| BC063234.1 | - | 10 - 11 | 1185 | 78 | ||
Set Membership: |
|