Search
Search

nucb2b| Gene Symbol: nucb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201479.1 | NP_958887.1 | 8 - 9 | 888 | 79 | |
| GenBank mRNA | BC047852.1 | - | 8 - 9 | 888 | 79 | |
| BC067334.1 | - | 8 - 9 | 878 | 79 | ||
Set Membership: |
|