Search
Search

nrp2b| Gene Symbol: nrp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212966.1 | NP_998131.1 | 7 - 8 | 1729 | 127 | |
| XM_017357663.2 | XP_017213152.1 | 7 - 8 | 1858 | 127 | ||
| GenBank mRNA | AY351913.1 | - | 7 - 8 | 1226 | 127 | |
| AY428802.1 | - | 7 - 8 | 1729 | 127 | ||
| AY437404.1 | - | 7 - 8 | 1689 | 127 | ||
| BC163576.1 | - | 7 - 8 | 1544 | 127 | ||
| BC163589.1 | - | 7 - 8 | 1544 | 127 | ||
| KM655620.1 | - | 7 - 8 | 1139 | 127 | ||
Set Membership: |
|