Search
Search

stx12| Gene Symbol: stx12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002051.1 | NP_001002051.1 | 8 - 9 | 757 | 73 | |
| XM_009294127.3 | XP_009292402.1 | 8 - 9 | 812 | 73 | ||
| XM_009294128.2 | XP_009292403.1 | 8 - 9 | 760 | 73 | ||
| GenBank mRNA | BC071327.1 | - | 8 - 9 | 757 | 73 | |
Set Membership: |
|