Search
Search

gla| Gene Symbol: gla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006103.2 | NP_001006103.2 | 5 - 6 | 837 | 95 | |
| GenBank mRNA | BC083209.1 | - | 5 - 6 | 838 | 95 | |
| BC164755.1 | - | 4 - 5 | 586 | 95 | ||
Set Membership: |
|