Search
Search

znf281b| Gene Symbol: znf281b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012487.2 | NP_001012505.2 | 3 - 4 | 757 | 132 | |
| XM_017358452.2 | XP_017213941.1 | 3 - 4 | 749 | 132 | ||
| GenBank mRNA | BC090180.1 | - | 3 - 4 | 761 | 132 | |
Set Membership: |
|