Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: clk4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013294.3 | NP_001013312.2 | 3 - 4 | 668 | 65 | |
NM_001329908.1 | NP_001316837.1 | 7 - 8 | 922 | 65 | ||
XM_009295587.3 | XP_009293862.1 | 4 - 5 | 485 | 65 | ||
GenBank mRNA | BC090270.1 | - | 3 - 4 | 595 | 65 | |
BC164422.1 | - | 1 - 2 | 189 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |