Search
Search

cbsb| Gene Symbol: cbsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014345.2 | NP_001014367.2 | 10 - 11 | 1324 | 109 | |
| GenBank mRNA | AY880251.1 | - | 10 - 11 | 1324 | 109 | |
Set Membership: |
|