Search
Search

erbb3b| Gene Symbol: erbb3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014826.1 | NP_001014826.1 | 18 - 19 | 2476 | 64 | |
| XM_005162170.4 | XP_005162227.1 | 19 - 20 | 2576 | 64 | ||
| XM_021469802.1 | XP_021325477.1 | 18 - 19 | 2568 | 64 | ||
| GenBank mRNA | AY863052.1 | - | 18 - 19 | 2476 | 64 | |
Set Membership: |
|